Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015305.3 | NP_056120.2 | 2 | 747 | |
XM_017021115.1 | XP_016876604.1 | 2 | 754 | ||
GenBank mRNA | AB018302.1 | - | 2 | 644 | |
AL137268.1 | - | 2 | 645 | ||
BC028714.2 | - | 2 | 694 | 4830897 |