Gene Transcripts

Gene Symbol
Rhox3
Entrez Gene ID
100190895
Gene Name
reproductive homeobox on X chromosome 3
Gene Aliases
Chromosome Location
Chr. X: 123862565 - 123868309 on Build Rnor_6.0
UniGene ID
Rn.18342
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135607.1 NP_001129079.1 435
XM_008773382.1 XP_008771604.1 236
XM_008773383.1 XP_008771605.1 3 182
XM_008773384.1 XP_008771606.1 121
GenBank mRNA DQ058651.1 - 435

Gene Symbol
LOC103690001
Entrez Gene ID
103690001
Gene Name
rhox homeobox family member 1-like
Gene Aliases
Chromosome Location
Chr. X: 124571933 - 124577677 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257477.2 XP_006257539.1 3 182
XM_006257478.2 XP_006257540.1 2 121
XM_008773510.1 XP_008771732.1 3 236

View Full Product Details