Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012089.2 | NP_036221.2 | 13 | 2102 | |
XM_011544135.2 | XP_011542437.1 | 13 | 2873 | ||
XM_011544136.2 | XP_011542438.1 | 11 | 1339 | ||
GenBank mRNA | AB013380.1 | - | 13 | 2089 | |
AF216833.1 | - | 13 | 2102 | ||
AK172794.1 | - | 12 | 1358 | ||
BC064930.1 | - | 13 | 2097 | 6143235 | |
DQ786241.1 | - | 6 | 480 | ||
U18237.1 | - | 6 | 532 |