Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012106.3 | NP_036238.1 | 3 | 371 | |
XM_011522977.1 | XP_011521279.1 | 3 | 110 | ||
GenBank mRNA | AF126062.1 | - | 3 | 230 | |
AK075050.1 | - | 3 | 371 | ||
AK298758.1 | - | 3 | 121 | ||
AK311018.1 | - | 3 | 263 | ||
BC003087.1 | - | 3 | 243 | 3503251 |