Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015500.1 | NP_056315.1 | 12 | 1690 | |
NM_199050.2 | NP_950251.1 | 11 | 1189 | ||
XM_005261109.4 | XP_005261166.1 | 11 | 1230 | ||
XM_011529522.1 | XP_011527824.1 | 12 | 1790 | ||
XM_011529523.2 | XP_011527825.1 | 13 | 2456 | ||
XM_011529524.2 | XP_011527826.1 | 12 | 1790 | ||
XM_017028315.1 | XP_016883804.1 | 9 | 926 | ||
GenBank mRNA | AB047784.1 | - | 12 | 1690 | |
AB047785.1 | - | 12 | 1690 | ||
AK125120.1 | - | 3 | 259 | ||
AK128550.1 | - | 9 | 1029 | ||
AL833323.1 | - | 11 | 1189 | ||
BC062323.1 | - | 11 | 1171 | 5419622 | |
CR627455.1 | - | 4 | 1464 |