Gene Transcripts

Gene Symbol
R3HCC1L
Entrez Gene ID
27291
Gene Name
R3H domain and coiled-coil containing 1 like
Gene Aliases
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location
Chr. 10: 98134608 - 98244897 on Build GRCh38
UniGene ID
Hs.419800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256619.1 NP_001243548.1 5 1396
NM_001256620.1 NP_001243549.1 4 1345
NM_014472.4 NP_055287.4 4 1341
NM_138469.2 NP_612478.2 5 1438
XM_011539639.1 XP_011537941.1 5 1653
XM_011539640.1 XP_011537942.1 6 1458
XM_011539641.1 XP_011537943.1 5 1410
XM_011539642.1 XP_011537944.1 6 1750
XM_011539643.1 XP_011537945.1 4 1337
XM_011539644.1 XP_011537946.1 4 1341
XM_011539645.2 XP_011537947.1 4 1584
XM_011539646.1 XP_011537948.1 3 1216
XM_011539647.1 XP_011537949.1 3 1218
XM_011539648.1 XP_011537950.1 2 1091
XM_011539649.1 XP_011537951.1 5 1653
XM_017016074.1 XP_016871563.1 4 2549
XM_017016075.1 XP_016871564.1 2 1091
XM_017016076.1 XP_016871565.1 5 1385
XM_017016077.1 XP_016871566.1 7 1548
XM_017016078.1 XP_016871567.1 6 1470
XM_017016079.1 XP_016871568.1 6 1484
XM_017016080.1 XP_016871569.1 4 1390
XM_017016081.1 XP_016871570.1 5 1472
XM_017016082.1 XP_016871571.1 3 1264
XM_017016083.1 XP_016871572.1 4 1280
GenBank mRNA AF050198.1 - 4 1332
AF525304.1 - 5 1349
AK292546.1 - 4 1340
BC098268.1 - 5 1379 40026913
BC098359.1 - 4 1282 40026912
BC099848.1 - 4 1282 40026916

View Full Product Details