Gene Transcripts

Gene Symbol
R3HCC1L
Entrez Gene ID
27291
Gene Name
R3H domain and coiled-coil containing 1 like
Gene Aliases
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location
Chr. 10: 98134608 - 98244897 on Build GRCh38
UniGene ID
Hs.419800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256619.1 NP_001243548.1 5 1570
NM_001256620.1 NP_001243549.1 4 1519
NM_014472.4 NP_055287.4 4 1515
NM_138469.2 NP_612478.2 5 1612
XM_011539639.1 XP_011537941.1 5 1827
XM_011539640.1 XP_011537942.1 6 1632
XM_011539641.1 XP_011537943.1 5 1584
XM_011539642.1 XP_011537944.1 6 1924
XM_011539643.1 XP_011537945.1 4 1511
XM_011539644.1 XP_011537946.1 4 1515
XM_011539645.2 XP_011537947.1 4 1758
XM_011539646.1 XP_011537948.1 3 1390
XM_011539647.1 XP_011537949.1 3 1392
XM_011539648.1 XP_011537950.1 2 1265
XM_011539649.1 XP_011537951.1 5 1827
XM_017016074.1 XP_016871563.1 4 2723
XM_017016075.1 XP_016871564.1 2 1265
XM_017016076.1 XP_016871565.1 5 1559
XM_017016077.1 XP_016871566.1 7 1722
XM_017016078.1 XP_016871567.1 6 1644
XM_017016079.1 XP_016871568.1 6 1658
XM_017016080.1 XP_016871569.1 4 1564
XM_017016081.1 XP_016871570.1 5 1646
XM_017016082.1 XP_016871571.1 3 1438
XM_017016083.1 XP_016871572.1 4 1454
GenBank mRNA AF525304.1 - 5 1523
AK094479.1 - 1 26
AK292546.1 - 4 1514
BC098268.1 - 5 1553 40026913
BC098359.1 - 4 1456 40026912
BC099848.1 - 4 1456 40026916

View Full Product Details