Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014008.4 | NP_054727.1 | 15 | 1843 | |
XM_005272599.3 | XP_005272656.1 | 15 | 1772 | ||
GenBank mRNA | AJ005890.1 | - | 15 | 1759 | |
AK291976.1 | - | 15 | 1804 | ||
AK298264.1 | - | 15 | 1838 | ||
AK316375.1 | - | 15 | 1789 | ||
BC000972.2 | - | 15 | 1790 | 3449051 | |
BC011675.2 | - | 15 | 1674 | 4299954 |