Gene Transcripts

Gene Symbol
C11orf21
Entrez Gene ID
29125
Gene Name
chromosome 11 open reading frame 21
Gene Aliases
Chromosome Location
Chr. 11: 2296277 - 2301913 on Build GRCh38
UniGene ID
Hs.559181
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142946.1 NP_001136418.1 1, 2 160
XM_011520034.1 XP_011518336.1 1, 2 160
GenBank mRNA AK310172.1 - 1, 2 160

View Full Product Details