Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013353.2 | NP_037485.2 | 7 | 824 | |
XM_011509449.1 | XP_011507751.1 | 7 | 706 | ||
XM_017001089.1 | XP_016856578.1 | 6 | 1441 | ||
GenBank mRNA | AF165217.1 | - | 7 | 701 | |
AF177173.2 | - | 7 | 769 | ||
AK092700.1 | - | 7 | 690 | ||
AK300675.1 | - | 5 | 527 | ||
AK313280.1 | - | 7 | 733 | ||
BC017810.1 | - | 7 | 734 | 4288769 |