Gene Transcripts
- Gene Symbol
- ALG5
- Entrez Gene ID
- 29880
- Gene Name
- ALG5, dolichyl-phosphate beta-glucosyltransferase
- Gene Aliases
- bA421P11.2
- Chromosome Location
- Chr. 13: 36949770 - 36999367 on Build GRCh38
- UniGene ID
- Hs.507769
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142364.1 |
NP_001135836.1
|
8, 9 |
844 |
|
NM_013338.4 |
NP_037470.1
|
9, 10 |
934 |
|
XM_017020552.1 |
XP_016876041.1
|
8, 9 |
872 |
|
GenBank mRNA |
AF088028.1 |
- |
4, 5 |
372 |
|
AF102850.1 |
- |
9, 10 |
942 |
|
AF161498.1 |
- |
9, 10 |
883 |
|
AF183413.1 |
- |
9, 10 |
893 |
|
AK299085.1 |
- |
8, 9 |
820 |
|
AK299126.1 |
- |
10, 11 |
1039 |
|
BC012531.1 |
- |
9, 10 |
886 |
4273297 |
View Full Product Details