Gene Transcripts

Gene Symbol
FAM216A
Entrez Gene ID
29902
Gene Name
family with sequence similarity 216 member A
Gene Aliases
C12orf24, HSU79274
Chromosome Location
Chr. 12: 110468424 - 110490387 on Build GRCh38
UniGene ID
Hs.436618
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013300.2 NP_037432.2 5 919
XM_005253875.4 XP_005253932.1 5 429
XM_011538246.2 XP_011536548.1 5 447
GenBank mRNA AK225731.1 - 5 910
AK296280.1 - 5 931
AK297684.1 - 5 429
AK308802.1 - 5 532
AK309603.1 - 4 443
BC020967.1 - 5 735 3848037
CR457001.1 - 5 470
U79274.1 - 5 886

View Full Product Details