Gene Transcripts

Gene Symbol
BFAR
Entrez Gene ID
51283
Gene Name
bifunctional apoptosis regulator
Gene Aliases
BAR, RNF47
Chromosome Location
Chr. 16: 14632811 - 14669236 on Build GRCh38
UniGene ID
Hs.435556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016561.2 NP_057645.1 7 1431
XM_005255350.2 XP_005255407.1 6 1206
XM_011522520.1 XP_011520822.1 5 1036
GenBank mRNA AF173003.1 - 7 1303
AK291114.1 - 7 1239
AK300778.1 - 5 1004
AK313890.1 - 6 1239
BC003054.1 - 7 1262 3537783

View Full Product Details