Gene Transcripts
- Gene Symbol
- ZDHHC3
- Entrez Gene ID
- 51304
- Gene Name
- zinc finger DHHC-type containing 3
- Gene Aliases
- DHHC-3, GODZ, ZNF373
- Chromosome Location
- Chr. 3: 44915261 - 44976190 on Build GRCh38
- UniGene ID
- Hs.61430
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135179.1 |
NP_001128651.1
|
6, 7 |
1024 |
|
NM_016598.2 |
NP_057682.1
|
7, 8 |
1108 |
|
XM_006713192.2 |
XP_006713255.1
|
8, 9 |
1210 |
|
XM_006713193.2 |
XP_006713256.1
|
7, 8 |
1127 |
|
XM_017006566.1 |
XP_016862055.1
|
7, 8 |
1062 |
|
XM_017006567.1 |
XP_016862056.1
|
7, 8 |
1100 |
|
XM_017006569.1 |
XP_016862058.1
|
7, 8 |
1099 |
|
XM_017006571.1 |
XP_016862060.1
|
7, 8 |
1104 |
|
GenBank mRNA |
AF247703.1 |
- |
7, 8 |
1048 |
|
AF441791.1 |
- |
5, 6 |
750 |
|
AY605661.1 |
- |
5, 6 |
528 |
|
BC015467.1 |
- |
6, 7 |
951 |
3852521 |
View Full Product Details