Gene Transcripts
- Gene Symbol
- SARAF
- Entrez Gene ID
- 51669
- Gene Name
- store-operated calcium entry associated regulatory factor
- Gene Aliases
- FOAP-7, HSPC035, TMEM66, XTP3
- Chromosome Location
- Chr. 8: 30063012 - 30083458 on Build GRCh38
- UniGene ID
- Hs.521487
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016127.5 |
NP_057211.4
|
2, 3 |
541 |
|
XM_017013567.1 |
XP_016869056.1
|
2, 3 |
791 |
|
XM_017013568.1 |
XP_016869057.1
|
2, 3 |
219 |
|
XM_017013569.1 |
XP_016869058.1
|
2, 3 |
219 |
|
GenBank mRNA |
AB028926.1 |
- |
2, 3 |
401 |
|
AF078855.1 |
- |
2, 3 |
369 |
|
AF100748.1 |
- |
2, 3 |
298 |
|
AF490252.1 |
- |
2, 3 |
282 |
|
AK025927.1 |
- |
2, 3 |
451 |
|
AK075340.1 |
- |
2, 3 |
399 |
|
AK094386.1 |
- |
3, 4 |
382 |
|
AK130057.1 |
- |
1, 2 |
149 |
|
AK222632.1 |
- |
2, 3 |
298 |
|
AK295360.1 |
- |
3, 4 |
346 |
|
AK314659.1 |
- |
2, 3 |
424 |
|
AY359104.1 |
- |
2, 3 |
466 |
|
BC015012.1 |
- |
2, 3 |
370 |
3908758 |
DC359554.1 |
- |
|
542 |
|
JQ348891.1 |
- |
2, 3 |
410 |
|
View Full Product Details