Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001178133.1 | NP_001171604.1 | 8 | 1143 | |
NM_016169.3 | NP_057253.2 | 8 | 1143 | ||
XM_011539858.2 | XP_011538160.1 | 8 | 1146 | ||
XM_011539860.2 | XP_011538162.1 | 8 | 1143 | ||
XM_011539861.2 | XP_011538163.1 | 8 | 1146 | ||
XM_011539863.2 | XP_011538165.1 | 8 | 881 | ||
XM_011539864.2 | XP_011538166.1 | 8 | 1146 | ||
XM_017016323.1 | XP_016871812.1 | 8 | 1376 | ||
GenBank mRNA | AF144231.1 | - | 8 | 1098 | |
AF159447.1 | - | 8 | 1098 | ||
AF172319.1 | - | 8 | 948 | ||
AF175770.1 | - | 8 | 1118 | ||
AF222345.1 | - | 8 | 1026 | ||
AK307370.1 | - | 3 | 352 | ||
AK311503.1 | - | 3 | 321 | ||
AK312319.1 | - | 8 | 1133 | ||
AY358550.1 | - | 8 | 1098 | ||
BC013291.2 | - | 8 | 1119 | 3533158 |