Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016653.2 | NP_057737.2 | 8 | 798 | |
NM_133646.2 | NP_598407.1 | 8 | 798 | ||
XM_005246640.2 | XP_005246697.1 | 8 | 844 | ||
XM_017004323.1 | XP_016859812.1 | 8 | 844 | ||
XM_017004324.1 | XP_016859813.1 | 8 | 868 | ||
GenBank mRNA | AB030034.1 | - | 8 | 773 | |
AB041883.1 | - | 8 | 868 | ||
AB049733.1 | - | 7 | 598 | ||
AB049734.1 | - | 7 | 598 | ||
AB208974.1 | - | 8 | 734 | ||
AF238255.1 | - | 7 | 598 | ||
AF251441.1 | - | 8 | 676 | ||
AF325454.1 | - | 7 | 603 | ||
AF465843.1 | - | 8 | 767 | ||
AF480461.1 | - | 8 | 793 | ||
AF480462.1 | - | 8 | 793 | ||
AK056310.1 | - | 8 | 799 | ||
AK291825.1 | - | 8 | 690 | ||
AK298634.1 | - | 7 | 563 | ||
BC001401.2 | - | 8 | 728 | 3051317 |