Gene Transcripts
- Gene Symbol
- CBSL
- Entrez Gene ID
- 102724560
- Gene Name
- cystathionine-beta-synthase like
- Gene Aliases
- Chromosome Location
- Chr. 21: 6444868 - 6468606 on Build GRCh38
- UniGene ID
- Hs.533013
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321073.1 |
NP_001308002.1
|
16, 17 |
1800 |
|
XM_011546099.1 |
XP_011544401.1
|
16, 17 |
1692 |
|
XM_017028209.1 |
XP_016883698.1
|
14, 15 |
1897 |
|
XM_017028210.1 |
XP_016883699.1
|
14, 15 |
1897 |
|
XM_017028214.1 |
XP_016883703.1
|
17, 18 |
2194 |
|
XM_017028215.1 |
XP_016883704.1
|
17, 18 |
1809 |
|
XM_017028216.1 |
XP_016883705.1
|
16, 17 |
2199 |
|
XM_017028217.1 |
XP_016883706.1
|
16, 17 |
1692 |
|
XM_017028218.1 |
XP_016883707.1
|
17, 18 |
1861 |
|
GenBank mRNA |
BC007257.1 |
- |
16, 17 |
1770 |
|
- Gene Symbol
- CBS
- Entrez Gene ID
- 875
- Gene Name
- cystathionine-beta-synthase
- Gene Aliases
- HIP4
- Chromosome Location
- Chr. 21: 43053190 - 43076868 on Build GRCh38
- UniGene ID
- Hs.533013
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000071.2 |
NP_000062.1
|
16, 17 |
1793 |
|
NM_001178008.2 |
NP_001171479.1
|
16, 17 |
1800 |
|
NM_001178009.2 |
NP_001171480.1
|
16, 17 |
1800 |
|
NM_001320298.1 |
NP_001307227.1
|
17, 18 |
1794 |
|
NM_001321072.1 |
NP_001308001.1
|
13, 14 |
1859 |
|
XM_011529775.2 |
XP_011528077.1
|
14, 15 |
1919 |
|
XM_011529776.2 |
XP_011528078.1
|
14, 15 |
1919 |
|
XM_017028490.1 |
XP_016883979.1
|
17, 18 |
2193 |
|
XM_017028491.1 |
XP_016883980.1
|
16, 17 |
1736 |
|
XM_017028492.1 |
XP_016883981.1
|
17, 18 |
1801 |
|
GenBank mRNA |
AK294608.1 |
- |
18, 19 |
1665 |
|
AK301063.1 |
- |
14, 15 |
1395 |
|
AK313691.1 |
- |
16, 17 |
1698 |
|
AL137314.1 |
- |
13, 14 |
1320 |
|
BC000440.2 |
- |
16, 17 |
1668 |
2820694 |
BC010242.1 |
- |
16, 17 |
1717 |
4154798 |
BC011381.1 |
- |
16, 17 |
1656 |
3869615 |
BC012319.1 |
- |
5, 6 |
470 |
|
BT007154.1 |
- |
14, 15 |
1548 |
|
L00972.1 |
- |
13, 14 |
1871 |
|
L14577.1 |
- |
16, 17 |
1706 |
|
L19501.1 |
- |
16, 17 |
1729 |
|
X82166.1 |
- |
16, 17 |
1695 |
|
View Full Product Details