Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017681.2 | NP_060151.2 | 4 | 362 | |
NR_033676.1 | 3 | 318 | |||
GenBank mRNA | AK000137.1 | - | 4 | 236 | |
BC016327.1 | - | 4 | 359 | 4102213 | |
BC017799.1 | - | 3 | 311 | ||
DB448067.1 | - | 3 | 319 | ||
KU178655.1 | - | 2 | 103 | ||
KU178656.1 | - | 2 | 103 |