Gene Transcripts
- Gene Symbol
- BBS7
- Entrez Gene ID
- 55212
- Gene Name
- Bardet-Biedl syndrome 7
- Gene Aliases
- BBS2L1
- Chromosome Location
- Chr. 4: 121824329 - 121870520 on Build GRCh38
- UniGene ID
- Hs.591694
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018190.3 |
NP_060660.2
|
16, 17 |
1965 |
|
NM_176824.2 |
NP_789794.1
|
16, 17 |
1965 |
|
XM_005263106.3 |
XP_005263163.1
|
16, 17 |
1991 |
|
XM_011532079.2 |
XP_011530381.1
|
16, 17 |
2036 |
|
XM_011532080.2 |
XP_011530382.1
|
16, 17 |
2033 |
|
XM_011532081.2 |
XP_011530383.1
|
15, 16 |
1871 |
|
XM_017008357.1 |
XP_016863846.1
|
15, 16 |
1823 |
|
XM_017008358.1 |
XP_016863847.1
|
15, 16 |
1826 |
|
GenBank mRNA |
AF521643.1 |
- |
16, 17 |
1929 |
|
AF521644.1 |
- |
16, 17 |
1929 |
|
AK001577.1 |
- |
16, 17 |
1929 |
|
AK308076.1 |
- |
16, 17 |
1954 |
|
BC032691.1 |
- |
16, 17 |
1882 |
5240398 |
View Full Product Details