Gene Transcripts

Gene Symbol
RHOT1
Entrez Gene ID
55288
Gene Name
ras homolog family member T1
Gene Aliases
ARHT1, MIRO-1, MIRO1
Chromosome Location
Chr. 17: 32142454 - 32253374 on Build GRCh38
UniGene ID
Hs.655325
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033566.2 NP_001028738.1 3, 4 409
NM_001033567.2 NP_001028739.2 4, 5 533
NM_001033568.2 NP_001028740.1 3, 4 409
NM_001288754.1 NP_001275683.1 3, 4 409
NM_001288755.1 NP_001275684.1 3, 4 405
NM_001288758.1 NP_001275687.1 4, 5 533
NM_018307.4 NP_060777.3 3, 4 409
NR_110083.1 3, 4 409
XM_011524969.2 XP_011523271.1 2, 3 133
XM_011524971.2 XP_011523273.2 3, 4 205
XM_011524973.2 XP_011523275.2 3, 4 205
XM_011524975.2 XP_011523277.1 4, 5 662
XM_011524976.2 XP_011523278.2 3, 4 205
XM_017024819.1 XP_016880308.1 4, 5 437
XM_017024820.1 XP_016880309.1 2, 3 133
XM_017024821.1 XP_016880310.1 3, 4 205
XM_017024822.1 XP_016880311.1 3, 4 434
XM_017024823.1 XP_016880312.1 3, 4 205
XM_017024824.1 XP_016880313.1 4, 5 540
XM_017024825.1 XP_016880314.1 4, 5 632
GenBank mRNA AJ496730.1 - 3, 4 397
AJ517412.1 - 3, 4 170
AK294407.1 - 3, 4 183
AL136929.1 - 3, 4 233
AY094972.1 - 3, 4 286
BC041114.1 - 3, 4 343
BC051818.1 - 2, 3 120
BC060781.1 - 3, 4 409 4823115
BC068463.1 - 3, 4 297
BC125104.1 - 3, 4 238 40118340
BC125105.1 - 3, 4 238 40118341
CN352790.1 - 3, 4 366

View Full Product Details