Gene Transcripts

Gene Symbol
TMEM55A
Entrez Gene ID
55529
Gene Name
transmembrane protein 55A
Gene Aliases
Chromosome Location
Chr. 8: 90994271 - 91040975 on Build GRCh38
UniGene ID
Hs.202517
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018710.2 NP_061180.1 6 824
GenBank mRNA AK313783.1 - 6 737
AX775961.1 - 6 674
BC033892.1 - 6 674 4820065
CR749733.1 - 5 388

View Full Product Details