Gene Transcripts
- Gene Symbol
- ASAP3
- Entrez Gene ID
- 55616
- Gene Name
- ArfGAP with SH3 domain, ankyrin repeat and PH domain 3
- Gene Aliases
- ACAP4, CENTB6, DDEFL1, UPLC1
- Chromosome Location
- Chr. 1: 23428563 - 23484631 on Build GRCh38
- UniGene ID
- Hs.437379
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017707.3 |
NP_060177.2
|
4, 5 |
546 |
|
XM_011541755.1 |
XP_011540057.1
|
4, 5 |
480 |
|
XM_017001685.1 |
XP_016857174.1
|
4, 5 |
480 |
|
XM_017001686.1 |
XP_016857175.1
|
4, 5 |
480 |
|
XM_017001687.1 |
XP_016857176.1
|
4, 5 |
453 |
|
XM_017001688.1 |
XP_016857177.1
|
4, 5 |
442 |
|
XM_017001689.1 |
XP_016857178.1
|
4, 5 |
497 |
|
GenBank mRNA |
AB056749.1 |
- |
4, 5 |
467 |
|
AK092291.1 |
- |
4, 5 |
468 |
|
AK299359.1 |
- |
4, 5 |
442 |
|
BC023519.2 |
- |
5, 6 |
534 |
|
BC060786.1 |
- |
4, 5 |
494 |
5298355 |
View Full Product Details