Gene Transcripts
- Gene Symbol
- SCYL2
- Entrez Gene ID
- 55681
- Gene Name
- SCY1 like pseudokinase 2
- Gene Aliases
- CVAK104
- Chromosome Location
- Chr. 12: 100267137 - 100341724 on Build GRCh38
- UniGene ID
- Hs.506481
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317784.1 |
NP_001304713.1
|
6, 7 |
1139 |
|
NM_017988.5 |
NP_060458.3
|
6, 7 |
1291 |
|
XM_017019618.1 |
XP_016875107.1
|
6, 7 |
1139 |
|
XM_017019619.1 |
XP_016875108.1
|
6, 7 |
1291 |
|
XM_017019620.1 |
XP_016875109.1
|
7, 8 |
1278 |
|
GenBank mRNA |
AB037781.1 |
- |
3, 4 |
445 |
|
AK000936.1 |
- |
5, 6 |
897 |
|
AK024274.1 |
- |
2, 3 |
300 |
|
AK027551.1 |
- |
2, 3 |
357 |
|
AK125091.1 |
- |
6, 7 |
1276 |
|
AK292980.1 |
- |
6, 7 |
1065 |
|
BC063798.1 |
- |
6, 7 |
904 |
6473586 |
View Full Product Details