Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018047.2 | NP_060517.1 | 2, 3 | 224 | |
GenBank mRNA | AK001152.1 | - | 2, 3 | 179 | |
AK297019.1 | - | 2, 3 | 140 | ||
AL136933.1 | - | 2, 3 | 153 | ||
BC003402.1 | - | 2, 3 | 157 | 3449258 | |
DA534416.1 | - | 2, 3 | 225 |