Gene Transcripts

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
Chromosome Location
Chr. 5: 141374084 - 141512979 on Build GRCh38
UniGene ID
Hs.368160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018919.2 NP_061742.1 1 999
NM_032086.1 NP_114475.1 1 999
GenBank mRNA AB209178.1 - 1 1169
AF152326.1 - 1 999
AF152513.1 - 1 999
BC109240.1 - 1 199
BC136702.1 - 1 1090 9020692
BC136703.1 - 1 1090 9020693
BC146651.1 - 1 1158 40147340

View Full Product Details