Gene Transcripts

Gene Symbol
ANKRD7
Entrez Gene ID
56311
Gene Name
ankyrin repeat domain 7
Gene Aliases
TSA806
Chromosome Location
Chr. 7: 118224658 - 118242731 on Build GRCh38
UniGene ID
Hs.729340
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019644.3 NP_062618.2 4 663
GenBank mRNA AK302407.1 - 4 617
BC007020.2 - 2 197
BC032799.1 - 4 424 5170634
D78334.1 - 4 419

View Full Product Details