Gene Transcripts
- Gene Symbol
- MAP3K7CL
- Entrez Gene ID
- 56911
- Gene Name
- MAP3K7 C-terminal like
- Gene Aliases
- C21orf7, HC21ORF7, TAK1L, TAKL, TAKL-1, TAKL-2, TAKL-4
- Chromosome Location
- Chr. 21: 29077471 - 29175889 on Build GRCh38
- UniGene ID
- Hs.222802
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286617.1 |
NP_001273546.1
|
4, 5 |
518 |
|
NM_001286618.1 |
NP_001273547.1
|
4, 5 |
518 |
|
NM_001286622.1 |
NP_001273551.1
|
4, 5 |
518 |
|
NM_001286623.1 |
NP_001273552.1
|
5, 6 |
675 |
|
NM_001286634.1 |
NP_001273563.1
|
4, 5 |
518 |
|
NM_020152.3 |
NP_064537.1
|
6, 7 |
747 |
|
GenBank mRNA |
AF269161.1 |
- |
4, 5 |
356 |
|
AF269162.1 |
- |
4, 5 |
356 |
|
AF269163.1 |
- |
4, 5 |
356 |
|
AK294287.1 |
- |
4, 5 |
498 |
|
AY171599.2 |
- |
5, 6 |
504 |
|
EF219157.1 |
- |
5, 6 |
675 |
|
View Full Product Details