Gene Transcripts

Gene Symbol
NCLN
Entrez Gene ID
56926
Gene Name
nicalin
Gene Aliases
NET59
Chromosome Location
Chr. 19: 3185877 - 3209575 on Build GRCh38
UniGene ID
Hs.657032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321463.1 NP_001308392.1 10 1432
NM_020170.3 NP_064555.2 10 1432
GenBank mRNA AK125002.1 - 10 1488
AK172848.1 - 10 1379
AK314044.1 - 10 1367
AL136567.1 - 10 1263
BC003076.2 - 2 122
BC007275.1 - 4 408
BC008920.2 - 6 639
BC010064.2 - 10 1348 4100399
BC013283.2 - 10 1709 3959506
BC019091.2 - 10 1348 4895124
BC025926.1 - 10 1344 3509966
CR533566.1 - 10 1245

View Full Product Details