Gene Transcripts

Gene Symbol
WDR18
Entrez Gene ID
57418
Gene Name
WD repeat domain 18
Gene Aliases
Ipi3, R32184_1
Chromosome Location
Chr. 19: 982980 - 994570 on Build GRCh38
UniGene ID
Hs.325321
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024100.3 NP_077005.2 1 93
XM_017027027.1 XP_016882516.1 3 619
GenBank mRNA AL560525.3 - 1 63
BC000040.2 - 1 78 3507673
BC001648.2 - 1 73 2819452
CB991338.1 - 120

View Full Product Details