Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000093.4 | NP_000084.3 | 4 | 924 | |
NM_001278074.1 | NP_001265003.1 | 4 | 924 | ||
XM_017014266.1 | XP_016869755.1 | 4 | 939 | ||
GenBank mRNA | AB209864.1 | - | 3 | 403 | |
AB371583.1 | - | 4 | 924 | ||
AK057231.1 | - | 5 | 963 | ||
D90279.1 | - | 4 | 667 |