Gene Transcripts

Gene Symbol
AMIGO1
Entrez Gene ID
57463
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
ALI2, AMIGO, AMIGO-1
Chromosome Location
Chr. 1: 109504178 - 109509742 on Build GRCh38
UniGene ID
Hs.726479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020703.2 NP_065754.2 2 1519
XM_011541812.2 XP_011540114.1 2 1525
GenBank mRNA AB032989.1 - 1 971
AK295678.1 - 2 1498
AY237007.1 - 1 1137
BC040879.1 - 2 1487 5725643

View Full Product Details