Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017519.2 | NP_059989.2 | 7 | 2375 | |
NM_020732.3 | NP_065783.3 | 8 | 2414 | ||
XM_005267069.4 | XP_005267126.2 | 7 | 2450 | ||
XM_011535984.2 | XP_011534286.2 | 9 | 2532 | ||
XM_017011103.1 | XP_016866592.1 | 9 | 2532 | ||
XM_017011104.1 | XP_016866593.1 | 9 | 2531 | ||
XM_017011105.1 | XP_016866594.1 | 9 | 2532 | ||
XM_017011106.1 | XP_016866595.1 | 9 | 2531 | ||
XM_017011108.1 | XP_016866597.1 | 9 | 2539 | ||
XM_017011109.1 | XP_016866598.1 | 5 | 4888 | ||
GenBank mRNA | AF253515.1 | - | 7 | 1569 | |
AF259792.1 | - | 3 | 317 | ||
AF468300.1 | - | 8 | 944 | ||
AF521671.1 | - | 7 | 2162 | ||
AK025945.1 | - | 5 | 641 | ||
AK074180.1 | - | 5 | 525 |