Gene Transcripts

Gene Symbol
STARD9
Entrez Gene ID
57519
Gene Name
StAR related lipid transfer domain containing 9
Gene Aliases
KIF16A
Chromosome Location
Chr. 15: 42575646 - 42720998 on Build GRCh38
UniGene ID
Hs.122061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020759.2 NP_065810.2 23 4625
XM_011521831.2 XP_011520133.1 23 4638
XM_011521832.2 XP_011520134.1 23 4635
XM_011521833.2 XP_011520135.1 22 4605
XM_011521834.2 XP_011520136.1 22 4575
XM_011521835.2 XP_011520137.1 21 4497
XM_011521836.2 XP_011520138.1 23 4940
XM_011521837.2 XP_011520139.1 21 4407
XM_011521839.2 XP_011520141.1 10 3407
XM_017022439.1 XP_016877928.1 22 4560
XM_017022440.1 XP_016877929.1 22 4537
XM_017022441.1 XP_016877930.1 21 4856
XM_017022442.1 XP_016877931.1 16 3964
XM_017022443.1 XP_016877932.1 15 3948
GenBank mRNA AK025589.1 - 1 1478

View Full Product Details