Gene Transcripts

Gene Symbol
BAHCC1
Entrez Gene ID
57597
Gene Name
BAH domain and coiled-coil containing 1
Gene Aliases
BAHD2
Chromosome Location
Chr. 17: 81395431 - 81466332 on Build GRCh38
UniGene ID
Hs.514580
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291324.1 NP_001278253.1 11 3639
XM_011525063.2 XP_011523365.1 12 4031
XM_017024898.1 XP_016880387.1 11 3690
XM_017024899.1 XP_016880388.1 10 3596
GenBank mRNA AB040880.1 - 7 1390

View Full Product Details