Gene Transcripts
- Gene Symbol
- USP37
- Entrez Gene ID
- 57695
- Gene Name
- ubiquitin specific peptidase 37
- Gene Aliases
- Chromosome Location
- Chr. 2: 218450251 - 218568393 on Build GRCh38
- UniGene ID
- Hs.166068
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020935.2 |
NP_065986.2
|
4, 5 |
563 |
|
XM_005246720.2 |
XP_005246777.1
|
4, 5 |
635 |
|
XM_005246721.3 |
XP_005246778.1
|
4, 5 |
527 |
|
XM_005246722.3 |
XP_005246779.1
|
4, 5 |
567 |
|
XM_005246724.1 |
XP_005246781.1
|
4, 5 |
563 |
|
XM_011511538.2 |
XP_011509840.1
|
3, 4 |
508 |
|
XM_011511541.2 |
XP_011509843.1
|
4, 5 |
563 |
|
XM_017004577.1 |
XP_016860066.1
|
4, 5 |
521 |
|
GenBank mRNA |
AK095577.1 |
- |
4, 5 |
521 |
|
AK128583.1 |
- |
4, 5 |
567 |
|
AL832645.2 |
- |
4, 5 |
562 |
|
BC050461.1 |
- |
4, 5 |
532 |
|
BC133007.1 |
- |
4, 5 |
448 |
40147029 |
BC133009.1 |
- |
4, 5 |
493 |
40147031 |
BC144249.1 |
- |
4, 5 |
448 |
9052769 |
BC144252.1 |
- |
4, 5 |
488 |
9052772 |
BX538024.1 |
- |
4, 5 |
528 |
|
View Full Product Details