Gene Transcripts

Gene Symbol
LY6G6D
Entrez Gene ID
58530
Gene Name
lymphocyte antigen 6 complex, locus G6D
Gene Aliases
C6orf23, G6D, LY6-D, MEGT1, NG25
Chromosome Location
Chr. 6: 31715356 - 31717804 on Build GRCh38
UniGene ID
Hs.591792
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021246.2 NP_067069.2 3 190
GenBank mRNA AJ315535.1 - 3 190
AJ315536.1 - 2 281
AJ315537.1 - 3 311
BC125138.1 - 3 194 40119583

View Full Product Details