Gene Transcripts

Gene Symbol
RMND5A
Entrez Gene ID
64795
Gene Name
required for meiotic nuclear division 5 homolog A
Gene Aliases
CTLH, GID2, GID2A, RMD5, p44CTLH
Chromosome Location
Chr. 2: 86720291 - 86778041 on Build GRCh38
UniGene ID
Hs.75277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022780.3 NP_073617.1 5 924
GenBank mRNA BC012165.1 - 5 645
BC047668.1 - 5 854 6066798

View Full Product Details