Gene Transcripts
- Gene Symbol
- SPATS2
- Entrez Gene ID
- 65244
- Gene Name
- spermatogenesis associated serine rich 2
- Gene Aliases
- Nbla00526, P59SCR, SCR59, SPATA10
- Chromosome Location
- Chr. 12: 49366905 - 49527441 on Build GRCh38
- UniGene ID
- Hs.654826
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293285.1 |
NP_001280214.1
|
12, 13 |
2092 |
|
NM_001293286.1 |
NP_001280215.1
|
10, 11 |
1945 |
|
NM_023071.3 |
NP_075559.2
|
11, 12 |
2008 |
|
NR_120663.1 |
|
12, 13 |
2052 |
|
XM_011538690.2 |
XP_011536992.1
|
12, 13 |
1480 |
|
XM_011538691.2 |
XP_011536993.1
|
10, 11 |
1375 |
|
XM_017019840.1 |
XP_016875329.1
|
11, 12 |
1448 |
|
GenBank mRNA |
AB075515.1 |
- |
9, 10 |
1036 |
|
AK023179.1 |
- |
7, 8 |
917 |
|
AK023202.1 |
- |
11, 12 |
1407 |
|
AK024678.1 |
- |
3, 4 |
304 |
|
AK095582.1 |
- |
12, 13 |
2052 |
|
AK292334.1 |
- |
10, 11 |
1345 |
|
AL833614.1 |
- |
12, 13 |
1533 |
|
BC048299.1 |
- |
10, 11 |
1375 |
5520818 |
BX648916.1 |
- |
10, 11 |
1318 |
|
View Full Product Details