Gene Transcripts
- Gene Symbol
- KATNBL1
- Entrez Gene ID
- 79768
- Gene Name
- katanin regulatory subunit B1 like 1
- Gene Aliases
- C15orf29
- Chromosome Location
- Chr. 15: 34140674 - 34210146 on Build GRCh38
- UniGene ID
- Hs.633566
- Species
- Homo sapiens
- Gene Symbol
- KATNBL1P6
- Entrez Gene ID
- 729176
- Gene Name
- katanin regulatory subunit B1 like 1 pseudogene 6
- Gene Aliases
- Chromosome Location
- Chr. 6: 146801669 - 146803824 on Build GRCh38
- UniGene ID
- Hs.486708
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003954.1 |
|
1 |
848 |
|
GenBank mRNA |
BC067801.1 |
- |
1 |
848 |
|
View Full Product Details