Gene Transcripts

Gene Symbol
SLTM
Entrez Gene ID
79811
Gene Name
SAFB like transcription modulator
Gene Aliases
Met
Chromosome Location
Chr. 15: 58879045 - 58934313 on Build GRCh38
UniGene ID
Hs.512932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013843.2 NP_001013865.1 15 2005
NM_024755.3 NP_079031.2 15 2059
NR_135042.1 13 1392
NR_135043.1 13 1502
XM_006720686.3 XP_006720749.3 13 2450
XM_006720690.1 XP_006720753.1 11 1156
XM_011522022.1 XP_011520324.1 15 2717
XM_011522023.1 XP_011520325.1 15 2663
XM_011522024.1 XP_011520326.1 14 2567
XM_011522025.1 XP_011520327.1 14 2519
XM_011522026.2 XP_011520328.2 14 2498
XM_011522027.1 XP_011520329.1 13 2471
XM_011522028.1 XP_011520330.1 13 2417
XM_011522029.2 XP_011520331.1 15 1898
XM_011522030.2 XP_011520332.1 15 2717
XM_011522031.2 XP_011520333.1 12 1553
XM_017022576.1 XP_016878065.1 14 1855
XM_017022577.1 XP_016878066.1 14 2444
XM_017022578.1 XP_016878067.1 13 2396
XM_017022579.1 XP_016878068.1 15 2062
XM_017022580.1 XP_016878069.1 14 1516
GenBank mRNA AK023275.1 - 5 513
AK055195.1 - 13 1295
AK291423.1 - 13 1425
BC014944.1 - 8 862
BC046119.1 - 4 354
BC140851.1 - 15 2026 9021719

View Full Product Details