Gene Transcripts

Gene Symbol
CATSPERB
Entrez Gene ID
79820
Gene Name
cation channel sperm associated auxiliary subunit beta
Gene Aliases
C14orf161, CatSper(beta)
Chromosome Location
Chr. 14: 91580774 - 91732086 on Build GRCh38
UniGene ID
Hs.131755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024764.3 NP_079040.2 7, 8 781
GenBank mRNA AK126034.1 - 7, 8 765
BC126214.1 - 7, 8 735 8991930
BC126216.1 - 7, 8 735 8991932

View Full Product Details