Gene Transcripts

Gene Symbol
TREML2
Entrez Gene ID
79865
Gene Name
triggering receptor expressed on myeloid cells like 2
Gene Aliases
C6orf76, TLT-2, TLT2, dJ238O23.1
Chromosome Location
Chr. 6: 41189749 - 41201233 on Build GRCh38
UniGene ID
Hs.164797
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024807.3 NP_079083.2 2 510
GenBank mRNA AF534824.1 - 2 509
AK023755.1 - 2 509
AY358171.1 - 2 511
BC125079.1 - 2 509 40117711

View Full Product Details