Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024939.2 | NP_079215.2 | 1 | 347 | |
XM_005256153.4 | XP_005256210.1 | 1 | 289 | ||
XM_006721273.3 | XP_006721336.1 | 1 | 34 | ||
GenBank mRNA | AK025571.1 | - | 1 | 246 | |
BC030146.2 | - | 1 | 169 | 4810948 | |
BC069241.1 | - | 1 | 698 | ||
BU943912.1 | - | 1 | 181 | ||
LN680438.1 | - | 1 | 159 | ||
LN680439.1 | - | 1 | 159 |