Gene Transcripts

Gene Symbol
TRABD
Entrez Gene ID
80305
Gene Name
TraB domain containing
Gene Aliases
LP6054, PP2447
Chromosome Location
Chr. 22: 50185889 - 50199599 on Build GRCh38
UniGene ID
Hs.517731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320484.1 NP_001307413.1 10 1853
NM_001320485.1 NP_001307414.1 10 1841
NM_001320487.1 NP_001307416.1 10 1875
NM_001320488.1 NP_001307417.1 11 1275
NM_025204.3 NP_079480.2 10 1866
NR_135275.1 10 1837
XM_006724422.3 XP_006724485.1 11 1245
XM_006724423.3 XP_006724486.1 11 1238
XM_011530712.2 XP_011529014.2 11 1313
XM_011530713.2 XP_011529015.2 10 1911
XM_011530715.2 XP_011529017.2 11 1289
XM_011530716.2 XP_011529018.1 11 1276
XM_011530717.2 XP_011529019.1 11 1257
XM_011530718.2 XP_011529020.1 11 1236
XM_017028943.1 XP_016884432.1 11 1318
XM_017028944.1 XP_016884433.1 10 1953
GenBank mRNA AL449244.1 - 10 1847
AY203945.1 - 9 1913
BC093029.1 - 9 1939 30527560
BX389277.2 - 5 533

View Full Product Details