Gene Transcripts

Gene Symbol
SPHKAP
Entrez Gene ID
80309
Gene Name
SPHK1 interactor, AKAP domain containing
Gene Aliases
SKIP
Chromosome Location
Chr. 2: 227979954 - 228181999 on Build GRCh38
UniGene ID
Hs.436306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142644.1 NP_001136116.1 6 568
NM_030623.3 NP_085126.2 6 568
XM_005246870.4 XP_005246927.1 7 1040
XM_006712777.3 XP_006712840.1 7 1001
XM_006712778.3 XP_006712841.1 7 1000
XM_011511925.2 XP_011510227.1 8 1119
GenBank mRNA AK127071.1 - 6 619
BC104954.1 - 6 568 8143957
BC104956.1 - 6 568 8143959
BC143408.1 - 6 568
CR749494.1 - 6 550

View Full Product Details