Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030783.1 | NP_110410.1 | 5 | 633 | |
XM_005253153.1 | XP_005253210.1 | 5 | 614 | ||
XM_011520391.2 | XP_011518693.1 | 5 | 1251 | ||
XM_011520392.2 | XP_011518694.1 | 6 | 630 | ||
XM_011520393.2 | XP_011518695.1 | 4 | 348 | ||
XM_017018368.1 | XP_016873857.1 | 7 | 3025 | ||
XM_017018369.1 | XP_016873858.1 | 6 | 755 | ||
XM_017018370.1 | XP_016873859.1 | 4 | 430 | ||
XM_017018371.1 | XP_016873860.1 | 5 | 544 | ||
GenBank mRNA | AL834357.1 | - | 5 | 547 | |
BC001210.1 | - | 5 | 633 | 3503123 |