Gene Transcripts
- Gene Symbol
- C10orf11
- Entrez Gene ID
- 83938
- Gene Name
- chromosome 10 open reading frame 11
- Gene Aliases
- CDA017
- Chromosome Location
- Chr. 10: 75431646 - 76559220 on Build GRCh38
- UniGene ID
- Hs.118161
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001305581.1 |
NP_001292510.1
|
3, 4 |
329 |
|
NM_032024.4 |
NP_114413.1
|
2, 3 |
381 |
|
NR_131178.1 |
|
4, 5 |
604 |
|
XM_011540256.2 |
XP_011538558.1
|
3, 4 |
323 |
|
XM_011540257.2 |
XP_011538559.1
|
3, 4 |
323 |
|
XM_017016759.1 |
XP_016872248.1
|
3, 4 |
323 |
|
XM_017016760.1 |
XP_016872249.1
|
3, 4 |
323 |
|
XM_017016761.1 |
XP_016872250.1
|
3, 4 |
323 |
|
GenBank mRNA |
AF267860.1 |
- |
2, 3 |
382 |
|
BM720401.1 |
- |
2, 3 |
329 |
|
BX105111.1 |
- |
2, 3 |
222 |
|
CR933639.1 |
- |
4, 5 |
604 |
|
HY006640.1 |
- |
3, 4 |
320 |
|
View Full Product Details