Gene Transcripts

Gene Symbol
ZDHHC18
Entrez Gene ID
84243
Gene Name
zinc finger DHHC-type containing 18
Gene Aliases
DHHC-18, DHHC18
Chromosome Location
Chr. 1: 26826710 - 26855720 on Build GRCh38
UniGene ID
Hs.523710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032283.2 NP_115659.1 1 420
GenBank mRNA AK056427.1 - 1 172
AL512765.1 - 1 25
BC066776.1 - 1 420 30342061

View Full Product Details