Gene Transcripts

Gene Symbol
FAM220A
Entrez Gene ID
84792
Gene Name
family with sequence similarity 220 member A
Gene Aliases
ACPIN1, C7orf70, SIPAR
Chromosome Location
Chr. 7: 6329409 - 6348959 on Build GRCh38
UniGene ID
Hs.696146
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037163.1 NP_001032240.1 2 1061
GenBank mRNA AF533707.1 - 2 1056
BC006110.1 - 2 1061 3139887
BC142977.1 - 2 1077 8860408

View Full Product Details